Imprimer
Multi-Purpose Matte Polyester Labels for M710 and BMP71
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
173351 173381 173360
BRDY173351EA 58.7 EUR
BRDY173351 BRDY173381 BRDY173360
Multi-Purpose Matte Polyester Labels for M710 and BMP71
Étiquettes
B-459 polyester from Brady® is designed for applications such as topside of printed circuit boards and rating plates that use high quality barcodes and graphics.
  • Matte white polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Excellent resistance to most chemicals, oils, and water
  • Service temperature of −40 to +100 °C ( −40 to +212 °F)
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, general identification, and inventory and inspection labelling applications.
Brady Authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Certifications: UL recognised.