Imprimer
Multi-Purpose Matte Polypropylene Labels for M710 and BMP71
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
173260 173291
BRDY173260EA 54.3 EUR
BRDY173260 BRDY173291
Multi-Purpose Matte Polypropylene Labels for M710 and BMP71
Étiquettes
The B-529 polypropylene tape is cost-effective with an acrylic pressure sensitive adhesive.
  • Ideal for use indoors on smooth, flat surfaces
  • Gloss finish
  • Service temperature of −40 to +100 °C (−40 to +212 °F)
  • Not recommended for outdoor use
Ideal for facility identification, general identification, and safety identification applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Cdt: 1 roll per box.