Imprimer
Harsh Environment Multi-Purpose Clear Polyester Labels for M710 and BMP71
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
173233 173319 173297 173226 173418 173409 174641 173266
BRDY173233EA 70.1 EUR
BRDY173233 BRDY173319 BRDY173297 BRDY173226 BRDY173418 BRDY173409 BRDY174641 BRDY173266
Harsh Environment Multi-Purpose Clear Polyester Labels for M710 and BMP71
Étiquettes
Clear polyester (B-430) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
  • Glossy, clear labels are thermal transfer printable and feature a durable permanent acrylic adhesive
  • Excellent resistance to most chemicals, oils and water
  • Service temperature of −56 to +137 °C (−70 to +100 °F)
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory andEn savoir plus