Imprimer
Polyester Autoclave and Cryogenic Labels for M710
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
311984 311981 311990 311991 311982 311978
BRDY311984EA 54.8 EUR
BRDY311984 BRDY311981 BRDY311990 BRDY311991 BRDY311982 BRDY311978
Polyester Autoclave and Cryogenic Labels for M710
Étiquettes
FreezerBondz™ material is a low-profile label used on frozen surfaces or on room-temperature surfaces that will eventually be stored in Liquid Nitrogen (-196°C).
  • Can be applied to frozen surfaces, including glass and polypropylene stored in liquid nitrogen
  • Good print smudge resistance and solvent resistance
  • High/low-temperature performance range of −196 to +100 °C
  • Matte finish
Identify frosted, frozen, or room temperature vials (with overlap). Opaque enough to cover legally protected patient information for compliance with HIPAA requirements. Ideal for general identification and laboratory labelling applications.