Imprimer
Clear Polyester Asset Tracking Labels for M710, M610 and M611
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
312058 312115 312122 312109
BRDY312058EA 63.5 EUR
BRDY312058 BRDY312115 BRDY312122 BRDY312109
Clear Polyester Asset Tracking Labels for M710, M610 and M611
Étiquettes
Labels specifically designed to fit Laminat™ Carriers and Tags. Suitable for printing with the M610 and M611 Thermal Transfer Portable Printers.
  • High-tack permanent adhesive for stainless steel, powder coated metals, painted enamel, and plastics
  • Resists exposure to UV light, extreme temperatures, fuel and oil, degreasers, cleaners, and detergents
  • Suitable for all-weather applications
  • Gloss finish
Ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and inventory and inspection labelling.
Certifications: UL recognised. CSA accepted.