Imprimer
Tamper Resistant Metallised Vinyl Labels for M710, M610 and M611
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
173868 173745 173726 173819
BRDY173868EA 77.3 EUR
BRDY173868 BRDY173745 BRDY173726 BRDY173819
Tamper Resistant Metallised Vinyl Labels for M710, M610 and M611
Étiquettes
Printable, tamper-evident, metallised vinyl labels for the M610 and M611 Portable Printers.
  • The metalized vinyl (B-362) tamper evident material is ideal for rating and serial plates
  • Use caution when removing when removing from liner as material is fragile
  • Material has great fuel and oil resistance
  • Service temperature of −70 to 100 °C (−94 to 212 °F)
  • Gloss finish
Ideal for general identification, inventory and inspection labelling, and security identification applications.
A ribbon is required to print this material. Use part number M61-R6010 for the M610 label printer/BradyPrinter M611 mobile label printer or M7-R6000 for the M710 label printer.