Imprimer
Self-Extinguishing Tedlar Wrap Around Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174174 174259 173715 173916
BRDY174174EA 135 EUR
BRDY174174 BRDY174259 BRDY173715 BRDY173916
Self-Extinguishing Tedlar Wrap Around Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Labels
Durable B-437 Topcoated Polyvinylfluoride material is used most frequently in wire and cable identification applications where self-extinguishing features are required.
  • White labels feature a matte finish and are thermal transfer printable
  • Permanent acrylic adhesive
  • Excellent resistance to water, oil, and other common fluids
Ideal for circuit board labelling, component and equipment labelling, general identification, electrical labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certifications: UL approved. CSA accepted. MIL-M-87958 Pressure Sensitive Adhesive Wire or Cable Marker and Identification specification.