Imprimer
Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M710, BMP71 Printers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
173235 173355 173239 173264 173348 173475 173312 173341 174607 173402 173225 174554 173293 173232 173396 173431 173405 173428 174606 173376
BRDY173235EA 63 EUR
BRDY173235 BRDY173355 BRDY173239 BRDY173264 BRDY173348 BRDY173475 BRDY173312 BRDY173341 BRDY174607 BRDY173402 BRDY173225 BRDY174554 BRDY173293 BRDY173232 BRDY173396 BRDY173431 BRDY173405 BRDY173428 BRDY174606 BRDY173376
Repositionable Multi-Purpose Vinyl Cloth Labels B-498 CleanLift® for M710, BMP71 Printers
Étiquettes
Permanent or repositionable, once applied the B-498 CleanLift® Series Vinyl Cloth remains in place, until it must be changed.
  • Adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Labels are ideal for asset tracking, barcode labelling, circuit board labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6200.
Certifications: UL recognised.