Imprimer
ToughWash® Washdown Resistant Labels for M710 and BMP71
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
173460 173461 173474 173473
BRDY173460EA 186 EUR
BRDY173460 BRDY173461 BRDY173474 BRDY173473
ToughWash® Washdown Resistant Labels for M710 and BMP71
Labels
The B-855 material is engineered to withstand rigorous washdowns and is ideal for safety and facility identification labelling and signage in the most aggressive environments.
  • B-855 print on demand pressure sensitive adhesive product for the food and beverage processing industry
  • Withstands high pressure, heat, harsh chemicals, and numerous washdown cycles
  • Legible and long-lasting for even the toughest washdown cycles
  • Service temperature range of −40 to +110 °C (−40 to +230 °F)
  • Matte finish
Ideal for food and beverage process labelling, general identification, and safety identification applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Cdt: One roll per box.