Imprimer
Retro Reflective All-Weather Labels for M610, M611 and BMP61
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174117 174130 174128 174129 174118
BRDY174117EA 180 EUR
BRDY174117 BRDY174130 BRDY174128 BRDY174129 BRDY174118
Retro Reflective All-Weather Labels for M610, M611 and BMP61
Labels
Highly reflective B-584 material shines brightly when struck by light, which is ideal for applications where high visibility is critical.
  • Retro reflective sheeting and pressure sensitive acrylic adhesive meet Federal Spec L-S-300 for Reflectivity
  • Average outdoor durability is 4 to 6 years
  • Service temperature of −40 to +70 °C (−40 to +158 °F)
  • Rflective gloss finish
Labels are ideal for component and equipment labelling, facility identification, general identification, and safety identification applications. Perfect for labelling job site tools, hard hats, utility enclosures, poles and pedestals, rooftop equipment, racks, and shelves.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010.
Cdt: One tape per box.