Imprimer
Retro Reflective All-Weather Labels for M710 and BMP71
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174509 174508 174514 174515 174510 174513
BRDY174509EA 170 EUR
BRDY174509 BRDY174508 BRDY174514 BRDY174515 BRDY174510 BRDY174513
Retro Reflective All-Weather Labels for M710 and BMP71
Étiquettes
Highly reflective B-584 material shines brightly when struck by light, which is ideal for applications where high visibility is critical.
  • Retro reflective sheeting and pressure sensitive acrylic adhesive meet Federal Spec L-S-300 for Reflectivity
  • Average outdoor durability is 4 to 6 years
  • Service temperature of −40 to +70 °C (−40 to +158 °F)
  • Reflective gloss finish
Labels are ideal for component and equipment labelling, facility identification, general identification, and safety identification applications. Perfect for labelling job site tools, hard hats, utility enclosures, poles and pedestals, rooftop equipment, racks, and shelves.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Cdt: One roll per box.