Imprimer
Magnetic Stirring Bars
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0076800 0076900 0076600 0076700
KART0076800EA 63.8 EUR
KART0076800 KART0076900 KART0076600 KART0076700
Magnetic Stirring Bars
Agitateurs Barreaux magnétiques
Economical stirring bars designed for a wide variety of applications.
  • Stronger mixing action at lower speeds
  • Can be used as an addition to any standard magnetic stir bar range, creating a viable alternative
  • The sizes shown below are nominal, with ±5% tolerance in length and ±10% in diameter
Suitable for foodstuff.