Imprimer
Stainless Steel Cleaners
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
EDEL600ES EDELS600
MMMAEDEL600ESEA 24.4 EUR
MMMAEDEL600ES MMMAEDELS600
Stainless Steel Cleaners
Produits nettoyants Détergents et Nettoyants de Surfaces
3M™ Stainless steel cleaners are cleaning sprays that create an invisible protective film which slows down the build-up of dirt particles and keeps surfaces cleaner for longer. With a pleasant citrus aroma, they clean, polish, and restore a glossy sheen to stainless steel and other metal surfaces.
  • Polishes and protects all stainless steel surfaces in one step
  • Spray clings to vertical surfaces and leaves a light, non-greasy film that resist fingerprints and marking
  • Does not contain harmful chemicals and is safe to use in food preparation areas
  • Water-based with a pleasant citrus fragrance
Wipe away dirt, fingerprints, grease, and residues on stainless steel and other metal surfaces with 3M™ Stainless steel cleaners. This fast and efficient spray thoroughly cleans all surfaces. Safe to use in food preparation areas, making it ideal for commercial kitchens and food processing areas. Also useful for cleaning and polishing architectural metal components such as lifts, escalators, and handrails.
Empty containers can be disposed of in sanitary landfills in accordance with local regulations. NSF® certified for use in meat and poultry processing plants.