Imprimer
Polyester Tapes, 8901
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3962533 89015066
MMMA3962533EA 1910 EUR
MMMA3962533 MMMA89015066
Polyester Tapes, 8901
Rubans adhésifs
Thin and conformable, 3M™ polyester tape 8901 excels in high temperature masking operations such as powder coat painting. Coated with a silicone adhesive, this tape bonds to a variety of difficult surfaces. It is chemical-resistant and performs under a wide range of temperatures and environments.
  • Works in demanding conditions and high temperatures ranging from –50 to +204 °C (–60 to +400 °F)
  • Clean removal during high-temperature masking operations such as powder coating
  • Polyester film offers excellent thermal and chemical resistance while maintaining
En savoir plus