Imprimer
Cole-Parmer® Stuart BH-200 Dry Block Heaters
Fournisseur: Cole Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36610-23 36610-11 36610-31 36610-32 36610-13 36610-01 36610-21
STUA36610-23EA 1470 EUR
STUA36610-23 STUA36610-11 STUA36610-31 STUA36610-32 STUA36610-13 STUA36610-01 STUA36610-21
Cole-Parmer® Stuart BH-200 Dry Block Heaters
Chauffe-blocs Bains à sec numériques
Provides precise heating of tubes and microplates.
  • Hot warning light indicates when the block temperature is above 50 °C
Dry block heaters are designed for precise heating of test tubes, microcentrifuge tubes, cuvettes, and microtiter plates. The large thermal mass of the aluminium blocks provides very accurate and stable temperature control. The 'number of blocks accepted' indicates the number of single-width blocks accepted by each heater. A heater that accepts two single-width blocks will also accept one double-width block.
Informations de livraison: Block extraction tool and power cord.