Imprimer
Cole-Parmer® HM-200 Multi-Position Extraction Heaters
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
03076-01 03076-04 03076-07 03076-10 03076-25 03076-28 03076-31 03076-34
ELET03076-01EA 1830 EUR
ELET03076-01 ELET03076-04 ELET03076-07 ELET03076-10 ELET03076-25 ELET03076-28 ELET03076-31 ELET03076-34
Cole-Parmer® HM-200 Multi-Position Extraction Heaters
Appareils chauffants Chauffe-ballons
Allow liquids with different boiling points to be heated next to each other.
  • Vented airflow ensures the case remains 'cool to the touch'
  • Heat your solutions simultaneously in each position
  • Robust construction permits continuous operation of the mantles as required by routine extraction tests
Regulating the heat on these multi-position extraction mantles is simple using the built-in solid-state electronic controls. The units accept round-bottom flasks ranging from 100 to 1000 ml. Select an extraction heater depending on the size of your flask.
Extraction heaters feature an independent heater per mantle, allowing liquids with different boiling points to be heated alongside each other. Each heater has its own control dial and on/off indicator light, and can reach up to a maximum element temperature of 450 °C (842 °F).
The control panel is safely separated from the heating elements by a well ventilated air space. A quartz heating element is housed in a thermally insulated cartridge for easy replacement, extending the life of the heater. Powder coated aluminium surfaces and polypropylene top cover ensure good chemical resistance and easy cleanup.
Informations de livraison: Supplied with two 71 cm long rods, one 140 cm long rod, support rod clamps, and 1,8 m power cord with EU-plug.