Imprimer
Cole-Parmer® HM-100 Uncontrolled Heating Mantles
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36003-06 36003-14 36003-18 36103-05
ELET36003-06EA 612 EUR
ELET36003-06 ELET36003-14 ELET36003-18 ELET36103-05
Cole-Parmer® HM-100 Uncontrolled Heating Mantles
Appareils chauffants Chauffe-ballons
Flexible heating element for efficient heat transfer and fast boiling times.
  • Require a power controller to operate
  • Exterior remains cool to the touch
  • Accepts round bottom flasks
  • Maximum element temperature of 450 °C (842 °F )
These simple to use, low-cost heating mantles quickly heat up to 450 °C (842 °F ). The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Powder-coated aluminium outer casing stays cool and is easy to clean. A stainless steel grounded earth screen and double fuses provide added safety. All heating mantles feature a support bracket for holding a support rod.
Informations de livraison: A controller is required to operate the heating mantle, this must be ordered separately.