Imprimer
Cole-Parmer® MSHM-200 Magnetic Stirring/Heating Mantles
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36013-00 36013-02 36013-04 36013-12
ELET36013-00EA 1370 EUR
ELET36013-00 ELET36013-02 ELET36013-04 ELET36013-12
Cole-Parmer® MSHM-200 Magnetic Stirring/Heating Mantles
Appareils chauffants Chauffe-ballons
Magnetic stirrer and heating mantle in one convenient unit.
  • Flexible coiled heating element wraps the heater around round-bottom flask to optimise heat transfer
  • 'Cool to the touch' exterior provides safe handling
  • Material: Aluminium
These stirring/heating mantles combine the functionality of a heating mantle with a magnetic stirrer providing even heating and mixing. Element temperatures can reach up to 450 °C (842 °F ) yet the exterior remains cool to the touch. For maximum safety, the flask does not come in direct contact with the heating element. Built-in energy regulator and safety earth (ground) screen offer added safety protection.
Reach stirring speeds up to 2000 rpm. Simply use the auto-recapture switch to reactivate stirring if stirring is interrupted and stirring motion is lost. Two coloured lights by each control dial indicate 'power on' and 'heat on'. Powder coated aluminium housing makes it easy to clean. All heating mantles feature a support bracket for holding a support rod.