Imprimer
WHEATON® CompletePAK® Sterile Components, Vials Only
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W987000-S W987001-S W987002-S W987000A-S W987001A-S W987002A-S
WHEAW987000-SEA 1070 EUR
WHEAW987000-S WHEAW987001-S WHEAW987002-S WHEAW987000A-S WHEAW987001A-S WHEAW987002A-S
WHEATON® CompletePAK® Sterile Components, Vials Only
Fioles
Clear or amber glass crimp finish vials sterilised, ready-to-use (RTU), complete manufacturing/certificate of analysis traceability included.
  • Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Elastomeric closures used in parenteral preparations
  • Complete manufacturing and certificate of analysis traceability
  • Ready-to-use, just-in-time, minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
  • Available in clear or amber glass for photosensitive contents
Certifications: USP 788 / EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers) USP 85 / EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers) and lot certified to SAL 10⁻⁶ (Vials, Stoppers, and Seals), USP 660 / EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381 / EP 3.2.9.