Imprimer
DURAN® Volumetric Flasks, Amber Glass, with PE Stopper
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
246740755 246740858 246741254 246741451 246741751 246742456 246743255 246743658 246744457 246745453 246746355
SCOT246740755EA 294 EUR
SCOT246740755 SCOT246740858 SCOT246741254 SCOT246741451 SCOT246741751 SCOT246742456 SCOT246743255 SCOT246743658 SCOT246744457 SCOT246745453 SCOT246746355
DURAN® Volumetric Flasks, Amber Glass, with PE Stopper
Ballons Fioles jaugées
Class A, delivered with UKAS certificate.
  • Made from borosilicate 3,3 glass
  • Each flask is individually tested and supplied with a calibration certificate complying to the requirements of ISO 17025 and is accredited at the United Kingdom Accreditation Service (UKAS)
  • The volume content tolerances conform to accuracy Class A. Calibration based on the poured in volume (‘In’) at a reference temperature of 20 °C
  • With etched graduation mark
  • UV protection due to external colouration, very durable and chemically resistant amber colour; the only external application of the colour ensures that internal surfaces have standard DURAN® glass properties
  • The white printed inscriptions are highly chemically resistant
DURAN® volumetric measuring instruments already meet the highest accuracy standard. The DURAN® range has now been expanded to include the product range of volumetric flasks with UKAS individual certificates. This new product line is tested and calibrated according to the requirements of the United Kingdom Accreditation Services (UKAS) and complies with ISO 17025, a standard for worldwide comparability of test and calibration results.
Certifications: Complies with ISO 1042.