Imprimer
Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
75003851 75003852 75003853 75003855
HERA75003851EA 316 EUR
HERA75003851 HERA75003852 HERA75003853 HERA75003855
Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Centrifugeuses Rotors
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
  • For high-throughput blood processing needs
  • Double quint blood bags (XXL, XL or M size)
  • Hook adapters for cord blood separations (XXL or M size)
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12×500 ml capacity or upgrade your centrifuge capacity to 16×500 ml blood bags with the HAEMAFlex 16 rotor and the same bucket selections.