Imprimer
Nunc™ Standard Closed Cell Factory Systems
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
140471 140472
734-3366EA 3570 EUR
734-3366 734-3367
Nunc™ Standard Closed Cell Factory Systems
Bioréacteurs
Advance your manufacturing process with Thermo Scientific™ Nunc™ Closed Cell Factory™ systems. Having a clean and closed system that provides consistent and high-quality results should be a standard among your adherent cell culture processes. This ready-to-use standard closed system will help you quickly develop and scale-up your process with standard components providing assurance of supply. Available in 2- and 10-tray versions.
  • Pre-configured closed system standard option
  • Sterility: Sterile
  • Makes it quick and easy to scale-up
  • Made to stock, Sartopore 0,2 µm vent filter
  • Aseptiquik G connector
  • Material: Polystyrene
  • Temperature range: 0 to 60 °C
  • Disposable
Ready-to-use out of the box.