Imprimer
Rubbermaid® Collapsible X-Carts
Fournisseur: Rubbermaid Commercial Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1889863 1889864 1871646 1871645 1871643 1871644
129-2070EA 77.4 EUR
129-2070 129-2063 129-2191 129-2193 129-2195 129-2194
Rubbermaid® Collapsible X-Carts
Chariots
Folding carts designed and tested to deliver reliable long-lasting performance.
  • Large capacity – up to 300 L capacity transports large loads easily, with smooth wheels that offer maximum manoeuvrability
  • Collapsible frame – facilitates efficient storage by reducing the space needed by up to 75%
  • Durable vinyl bag – tear-resistant and hard-wearing for heavy duty use
  • Silent operation – reduces disturbances in environments that are dependent on quiet; ideal for hospitality and medical settings
  • Discreet colour – blends into most environments and provides a clean, professional look
  • Multiple, built-in storage pockets – on each side keep frequently used cleaning tools and supplies within reach
  • Convenient, quick access lid – provides easy access to contents inside of cart without having to remove cover; secures with velco trim
Rubbermaid's X-Cart range consists of folding trolleys that are capable of transporting large loads without impacting on manoeuvrability. Their collapsible design means that they can be stored with ease, even in environments where space is an issue. Silent operation reduces disturbances in environments that are dependent on quiet.
Informations de commande: The new Rubbermaid® X-cart bags and frames are sold separately, which allows you to choose one of the following configurations:
150 L frame + 150 L bag;
300 L frame + 300 L bag;
or 300 L frame + 2 x 150 L bags (2-stream configuration for sorting, facilitated by colour ribbons to avoid cross-contamination)