Imprimer
Reaction Stations, RS-200 and RS-250
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
36630-01 36630-11
ELET36630-01EA 13890 EUR
ELET36630-01 ELET36630-11
Reaction Stations, RS-200 and RS-250
Agitateurs Agitateurs magnétiques chauffants
Run multiple reactions in a single station, saving time and money.
  • Control and maintain the same temperature and stirring rate at each position
  • Excellent temperature uniformity across the block
  • Simple menus and touchpad interface to control the system performance
  • Exterior remains cool to the touch for added safety
Run these reaction stations at the same temperature and stirring rates at several positions simultaneously. They are great for a wide range of applications from simple synthesis to process optimisation. High-performance magnetic stirrersEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Modèle RS-200 RS-250
Environmentally Preferable
Exigence électrique 230 VAC, 50 Hz 230 VAC, 50 Hz
Gamme de vitesse 100 - 800 min⁻¹ 400 - 2000 min⁻¹
L×P×H 23,5×33×30,2 cm 8×30,5×15 cm
Gamme de température 220 °C 150 °C