Imprimer
Masterflex® USB A Serial Adapter Cables, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX78006-90EA 131 EUR
MFLX78006-90 MFLX78006-91
Masterflex® USB A Serial Adapter Cables, Avantor®
Cables
Optimize performance of your Masterflex® MasterSense™ or Ismatec® Reglo Touchscreen pump.
  • Connect your drive via USB A for serial control
Not your standard USB cable. Use one of these cables to connect a Masterflex® MasterSense™ and Ismatec® Reglo Touchscreen pumps to a PC or PLC for serial (RS232) communicaiton. Cables features special capability to support serial interface communication. Chose USB A to USB A for communication to computers. The three wire cable is desinged for serial communcation with PLC applications.