Imprimer
BASE Footwear Grand Canyon/Tulum Safety Shoes, Lace-UP, Low, S1P, Grey
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B0666GRR35 B0666GRR36 B0666GRR37 B0666GRR38 B0666GRR39 B0666GRR40 B0666GRR41 B0666GRR42 B0666GRR43 B0666GRR44 B0666GRR45 B0666GRR46 B0666GRR47 B0666GRR48
PRTWB0666GRR35EA 138 EUR
PRTWB0666GRR35 PRTWB0666GRR36 PRTWB0666GRR37 PRTWB0666GRR38 PRTWB0666GRR39 PRTWB0666GRR40 PRTWB0666GRR41 PRTWB0666GRR42 PRTWB0666GRR43 PRTWB0666GRR44 PRTWB0666GRR45 PRTWB0666GRR46 PRTWB0666GRR47 PRTWB0666GRR48
BASE Footwear Grand Canyon/Tulum Safety Shoes, Lace-UP, Low, S1P, Grey
Chaussures et bottes
Grand Canyon/Tulum low safety shoes in grey feature a Microfibre + Hi-Tech fabric upper, SmellStop anti-bacterial lining, Fresh’n Flex midsole, SlimCap toe cap, and AirTech + TPU-skin sole.
  • 200 joules resistant toe-cap
  • Puncture-resistant sole
  • Shock absorption in the heel region
  • Anti-static footwear
  • Electronic devices protection from electrostatic discharges
  • Hydrocarbons-resistant
  • Metal-free
Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces. Orthopaedic made to measure footbed, complying with european norms DGUV112-191.
Certifications: EN ISO 20345: 2011 S1P FO ESD SR; CEI EN 61340-4-3 S1P.