Imprimer
MacCONKEY Agar acc. ISO 21150, ISO 21567, FDA-BAM and EP, USP, JP GranuCult® prime, Millipore
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.00205.0500 1.00205.5000
1.00205.0500EA 241 EUR
1.00205.0500 1.00205.5000
MacCONKEY Agar acc. ISO 21150, ISO 21567, FDA-BAM and EP, USP, JP GranuCult® prime, Millipore
Media for Microbiology Dehydrated Media for Microbiology
For the isolation of Salmonella, Shigella and coliform bacteria from foodstuffs and wastewater.