Imprimer
Core Inkjet Polypropylene Roll Labels, 76,20 mm
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
153635 153636 153651 153652 153653 153654 153656
BRDY153635EA 209 EUR
BRDY153635 BRDY153636 BRDY153651 BRDY153652 BRDY153653 BRDY153654 BRDY153656
Core Inkjet Polypropylene Roll Labels, 76,20 mm
Labels
This B-7425J polypropylene material is ideal for full-colour identification and barcode applications where resistance to liquid nitrogen, autoclave conditions, and harsh chemicals is required.
  • White polypropylene with acrylic adhesive is inkjet printable
  • Perfect for laboratory identification where labels may come in contact with harsh chemicals
  • Designed for roll inkjet printers
  • Service temperature of −80 to +70 °C (−112 to +158 °F)
  • Matte finish
Applications include barcode labelling, cold temperature labelling, COVID-19, general identification, hazard communication identification, and laboratory labelling.