Imprimer
Pipette holders, Rainin Hang-Up™
Fournisseur: RAININ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HU-M3 HU-S3
613-4724EA 175 EUR
613-4724 613-4725
Pipette holders, Rainin Hang-Up™
Portoirs pour pipettes
Holders for up to three manual, electronic or multi-channel pipettes. Available magnetic or with shelf bracket.
  • HU-M3: Magnetized for ferrous surfaces, adhesive disks supplied
  • HU-S3: Three holders mounted on a shelf bracket; quickly attaches to the edge of a shelf
Adapter 17006638 is needed for hanging electronic or multi-channel pipettes.