Imprimer
Separating Funnels, Squibb
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
632426355056 632426355106 632426355256 632426355506 632426355946
KAVA632426355056EA 125 EUR
KAVA632426355056 KAVA632426355106 KAVA632426355256 KAVA632426355506 KAVA632426355946
Separating Funnels, Squibb
Entonnoirs Ampoules à décanter
Borosilicate 3.3 glass, delivered with NS-PE stopper.
  • Very good chemical and temperature resistence
  • Stopcock with PTFE key
  • Without graduations
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable