Imprimer
Cole-Parmer® Heating Blocks and Heating Block Kits for Hotplates and Magnetic Hotplate Stirrers
Fournisseur: Cole Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
04805-76 04805-78 04805-79 04805-87 04805-85 04805-83 04805-81 04805-89 04805-97 04805-98
STUA04805-76EA 555 EUR
STUA04805-76 STUA04805-78 STUA04805-79 STUA04805-87 STUA04805-85 STUA04805-83 STUA04805-81 STUA04805-89 STUA04805-97 STUA04805-98
Cole-Parmer® Heating Blocks and Heating Block Kits for Hotplates and Magnetic Hotplate Stirrers
Chauffe-blocs Blocs pour bains
Turn an existing 15,2×15,2 cm hotplate into a heating mantle.
  • Safest way to heat round-bottom flasks
  • Flexible modular system for a wide range of sizes up to 500 ml
  • Low-well design is perfect for multi-neck flasks
Designed to heat round-bottom flasks, the heating block system with interchangeable inserts can be used with various Cole-Parmer hotplates and magnetic hotplate stirrers with a 6×6" top plate. The heating block system offers two sizes of base plates (small and medium) which can accommodate up to three block inserts each. A third larger base plate features heat-resistant handles and accepts a single block insert for flasks up to 500 ml.
Chemical- and solvent-resistant anodised block inserts provide a safe method for synthesis of single or multiple reactions and offer equal or better heating performance than silicon oil. The low-well provides excellent temperature transfer without compromising the view of the reaction or risk of cracking the glassware. Each base plate features a hole to house a temperature controller probe.