Imprimer
Cole-Parmer® Glass Capillary Tubes
Fournisseur: Cole Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
03013-63 03013-64 03013-65
STUA03013-63EA 35.5 EUR
STUA03013-63 STUA03013-64 STUA03013-65
Cole-Parmer® Glass Capillary Tubes
Tubes Capillaires de point de fusion
Accessories for your melting point apparatus.
  • Constructed of soda glass and easily sealed in a Bunsen flame
  • Supplied in a durable tube holder for added protection
Capillary tubes measure 100 mm L, 1,9 mm OD, 1,3 mm ID, and 0,3 mm wall thickness.