Imprimer
Cole-Parmer® IC-300 Series Digital Immersion Circulators
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
01262-07 16112-55
TECN01262-07EA 1880 EUR
TECN01262-07 TECN16112-55
Cole-Parmer® IC-300 Series Digital Immersion Circulators
Thermostats à circulation Systèmes à circulation ouverte Contrôleurs à thermostat
Accurate PID temperature control.
  • 4 digit setting with a bright LED digital temperature display
  • Suitable for most routine laboratory applications
  • User adjustable over-temperature cut-out
  • Low liquid level cut-out as standard
  • Temperature Sensor: 100 Ω Pt RTD
These versatile immersion circulators can be used in several ways. Use as a stand-alone water bath temperature controller, a pump for recirculating chilled liquid from an external chiller, or a pump for providing temperature control to an external device. The circulators feature a digital LEDEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Modèle IC-300-10DDC IC-300-20HDC
Environmentally Preferable
Max. pump pressure flow 10 LPM, 2,1 PSI 10 LPM, 2,1 PSI
Affichage LED LED
Alimentation 230 VAC, 50/60 Hz
Exigence électrique 230 VAC, 50/60 Hz
L×P×H 124×237×260 mm 124×237×260 mm