Imprimer
Polysorbate 80 FCC
Marques : Spectrum Chemical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
P1179-20LT P1179-4LTGL
SPCMP1179-20LTEA 1140 EUR
SPCMP1179-20LT SPCMP1179-4LTGL
Polysorbate 80 FCC
Polysorbate 80
Polysorbate 80, FCC is used as a wetting agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Kosher certified
Storage temperature - Keep tightly closed, protect from light.
Point de fusion: 100 °C
Point éclair: 113 °C (coupe fermée)
Température de stockage: Température ambiante
Numéro MDL: MFCD00082107
Numéro CAS: 9005-65-6
EINECS: 500-019-9
Commander maintenant
Résultats des tests de spécification
Assay 65% Min.