Polyethylene glycol 600 NF
Marques : Spectrum ChemicalTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Polyethylene Glycol 600, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formule:
H(OCH₂CH₂)nOH Point d'ébullition: ∼250 °C (1013 hPa) Point de fusion: 17…22 °C Densité: 1,128 Point éclair: 270 °C Température de stockage: Température ambiante |
Numéro MDL:
MFCD00081839 Numéro CAS: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...