Imprimer
Bole white (kaolin), en poudre FCC
Marques : Spectrum Chemical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): China Clay, Hydrated aluminum silicate, Clay
K1004-500GM K1004-2.5KG
SPCMK1004-500GMEA 145 EUR
SPCMK1004-500GM SPCMK1004-2.5KG
Bole white (kaolin), en poudre FCC
Kaolin blanc
Kaolin, Powder, FCC is used as a whitener in toothpaste. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: Al₂SiO₇·2H₂O
Poids moléculaire: 258.16 g/mol
Point de fusion: 1760
Température de stockage: Température ambiante
Numéro MDL: MFCD00062311
Numéro CAS: 1332-58-7
Indice Merck: 13,05300
Commander maintenant
Résultats des tests de spécification
Acid-Soluble Substances max. 2.0%
Arsenic (As) max.3 mg/kg
Carbonate (CO₃) To Pass Test
Iron (Fe) To Pass Test
Lead (Pb) max. 10 mg/kg