Imprimer

Calamine ≥98.0%, en poudre USP

Marques : Spectrum Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

CA115-12KG CA115-2.5KG CA115-45KGBL
SPCMCA115-12KGEA 1450 EUR
SPCMCA115-12KG SPCMCA115-2.5KG SPCMCA115-45KGBL
Calamine ≥98.0%, en poudre USP
Calamine

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Température de stockage: Température ambiante
Numéro CAS: 8011-96-9

Commander maintenant

Résultats des tests de spécification

Assay (ZnO) 98.0 - 100.5%
Microbial Limits To Pass Test
Loss On Ignition max. 2.0 %
Acid-Insoluble Substances max. 2.0 %
Alkaline Substances To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR