Imprimer

2-(Dimethylamino)-ethanol ≥99%

Marques : Spectrum Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Synonyme(s): DMAE

D2399-2.5LTGL D2399-200LTBL D2399-20LT
SPCMD2399-2.5LTGLEA 438 EUR
SPCMD2399-2.5LTGL SPCMD2399-200LTBL SPCMD2399-20LT
2-(Dimethylamino)-ethanol ≥99%
2-Diméthylaminoéthanol

N,N-Dimethylethanolamine is a transparent, pale yellow liquid and primary alcohol that is used as a building block for the synthesis of cationic flocculants and ion exchange resins. It is also used used as a chemical intermediate for pharmaceuticals, dyes, corrosion inhibitors, and emulsifiers and as an additive to boiler water, paint removers, and amino resins.


GHS hazard : Corrosion;Flame;Skull and crossbones;Exclamation mark
UN2051

Formule: C₄H₁₁NO
Poids moléculaire: 89.14 g/mol
Point de fusion: -59
Densité: 0,887
Point éclair: 39
Température de stockage: Température ambiante
Numéro MDL: MFCD00002846
Numéro CAS: 108-01-0
UN: 2051
ADR: 8.3 II
Indice Merck: 13,02863

Commander maintenant

Résultats des tests de spécification

Assay 0.99
Color (APHA) max. 20
Water max. 0.1 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR