Imprimer
Potassium carbonate, anhydre ≥99%, granuleux FCC
Marques : Spectrum Chemical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Carbonate de potassium
P1237-12KG P1237-2.5KG
SPCMP1237-12KGEA 992 EUR
SPCMP1237-12KG SPCMP1237-2.5KG
Potassium carbonate, anhydre ≥99%, granuleux FCC
Potassium carbonate
Potassium Carbonate, Anhydrous, Granular, FCC is used as a meat tenderizer, a source of dietary potassium and a buffering agent in wine production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formule: K₂CO₃
Poids moléculaire: 138.21 g/mol
Point de fusion: 891 °C
Densité: 2,42 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00011382
Numéro CAS: 584-08-7
EINECS: 209-529-3
Indice Merck: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM
Commander maintenant
Résultats des tests de spécification
Assay 99% Min.