Imprimer
Pre-wetted cleanroom wipes, ThermaSat™ 60
Fournisseur: Texwipe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TX4304 STX4305 STX4306 TX4307 TX4308
TEXWTX4304EA 278 EUR
TEXWTX4304 TEXWSTX4305 TEXWSTX4306 TEXWTX4307 TEXWTX4308
Pre-wetted cleanroom wipes, ThermaSat™ 60
Lingettes Lingettes pour salles propres Lingettes pour salles propres, présaturées
ThermaSat™ 60 wipes are made from 100% polyester with a sealed edge, and cleanroom laundered.
  • Wipe and clean surfaces, equipment, and parts
  • Apply and remove lubricants, adhesives, residues, and other solutions including disinfectants
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers
  • Clean gloves, notebooks, phones, or any other item entering the cleanroom
  • Line trays for holding, protecting, drying, and storing of parts, equipment, and devices
  • Appropriate for use with temperatures less than 205 °C (400 °F) (dry wipes only)
Constructed from 100% polyester to create a wipe ideal for spill control, cleaning, and solution application. ThermaSat™ wipes are packaged in easy-to-open bags (perforated bag, slider bag, or peel and reseal bag). They offer excellent chemical resistance for compatibility with a variety of solutions. ThermaSat™ wipes provide consistent, optimized cleaning efficiency with repeatable wetness and VOC levels. Individually lot coded for ease of traceability and quality control.
Certifications: Meets USP 797 and USP 800 wiper requirements.