Imprimer
Lecture Bottle (LB) Gas Cylinder Holder
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
35322 35316
JUSU35322EA 287 EUR
JUSU35322 JUSU35316
Lecture Bottle (LB) Gas Cylinder Holder
Stockage des fûts
Lecture Bottle (LB) Gas Cylinder Holder provides safety when moving and storing of gas cylinders.
  • Sized to fit 51 mm (2") calibration gas cylinders or LB style lecture bottles
  • Mount holes: 6,35 mm (0,25")
  • Durable and chemical resistant
  • Designed to either be free-standing or bench mounted with pre-drilled mount holes
The Carrier model is made from chemically welded PVC with formed walls and a loop handle.
Certifications: Holders are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA approved.