Imprimer
Cole-Parmer® Essentials Cell Culturing Flasks for BS-200 Series Cell Culture Stirring Systems
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
21837-37 21837-34 21837-30 21837-31
TECN21837-37EA 654 EUR
TECN21837-37 TECN21837-34 TECN21837-30 TECN21837-31
Cole-Parmer® Essentials Cell Culturing Flasks for BS-200 Series Cell Culture Stirring Systems
Ballons Flacons pour culture cellulaire
A complete culture vessel (flask, stir rod, and caps) to use with biological stirrers.
  • Completely sealed vessel is ideal for pathogenic materials
Culture vessels come calibrated and have two side ports and a central filling port. All culture vessel parts can be autoclaved at 121 °C and 15 psi for 30 minutes. Silicone-encapsulated stir rods reduce the possibility of cells attaching to and growing on the surface.