Imprimer
Cole-Parmer® BS-200 Cell Culture Stirring Systems (Stirrers)
Fournisseur: Cole-Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
21837-01 21837-02 21837-03 21837-04
TECN21837-01EA 3140 EUR
TECN21837-01 TECN21837-02 TECN21837-03 TECN21837-04
Cole-Parmer® BS-200 Cell Culture Stirring Systems (Stirrers)
Agitateurs Agitateurs magnétiques multipostes
Gentle stirring action promotes high cell yields.
  • Soft start/stop avoids excessive turbulence in the culture media
  • Interval stirring is ideal for extremely fragile cells
  • Temperature : 40 °C
The cell culture stirring system incorporates everything needed to achieve high cell yields for your application. The soft start/stop feature provides slow acceleration and deceleration of the stirrer, avoiding excessive turbulence in the culture media and eliminating cell damage. Use the interval stirring mode during the attachment phase to further reduce agitation of the media or when culturing particularly fragile cells. The stirrers create virtually no heat making them suitable for use in both incubators and cold rooms.
Use the biological stirrer with Cole-Parmer® Essentials culturing flasks—order separately. The culture flasks with their base design and the bulb-ended stirrer ensure that the cells are lifted into suspension at the lowest possible speeds promoting high cell yields.
Informations de livraison: Supplied with three power cords with plug (US, UK, and European).