Imprimer
ThermoSafe Large-Capacity Dry Ice Storage Chests
Fournisseur: Cole Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
03721-02 03720-00 03720-10 03721-00 03721-10
STUA03721-02EA 1430 EUR
STUA03721-02 STUA03720-00 STUA03720-10 STUA03721-00 STUA03721-10
ThermoSafe Large-Capacity Dry Ice Storage Chests
Refroidisseurs Refroidisseurs de paillasses
High capacity storage for transport of dry ice pellets and blocks.
  • Double-wall polyethylene construction filled with urethane foam insulation
  • Perfect for short or long-term storage
  • Ideal for dispensing dry ice in a laboratory environment
  • Special formulated insulation for lowest sublimation rates
Large-Capacity dry ice storage chests feature a lightweight construction with special formulated insulation for lowest sublimation rates. Chests contain extra-thick foam plastic insulation core.
Move the large-capacity storage chests easily on the aluminum dollies, even when filled to capacity with dry ice. The rim on the edge of platform keeps containers on the dolly during acceleration, bumps, or stops. Dollys feature a gray hammertone finish on heavy-gauge aluminum.
Sublimation rates: The 1,6 cu. ft. chest requires 6 days for 85 lbs. of dry ice to sublimate to 5 lbs. The 2,5 cu. ft. chest requires 10 days for 150 lbs. of dry ice to sublimate to 5 lbs. The 3,75 cu. ft. chest requires 15 days for 200 lbs. of dry ice to sublimate to 5 lbs.