Imprimer

Anti-COX4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BOSSBS-10257R-A750EA 356 EUR
BOSSBS-10257R-A750
Anti-COX4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticorps
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.

Type: Primary
Antigen: COX4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-169/169
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR