Imprimer

Anti-Tubulin-beta Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BOSSBS-0210R-A680EA 356 EUR
BOSSBS-0210R-A680
Anti-Tubulin-beta Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticorps
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.

Type: Primary
Antigen: Tubulin-beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-50/444
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR