Imprimer
Goat Serum Heat Inactivated, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
092939254 092939249
ICNA092939254EA 134 EUR
ICNA092939254 ICNA092939249
Goat Serum Heat Inactivated, MP Biomedicals
Sérums
Goat Serum is used as a supplement to cell culture media.
  • Collected from whole blood from healthy individuals
  • Heat inactivated
  • Liquid Media
  • Store at —20 °C
Goat Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health.
Be certain to maintain good cell culture practice, and maintain sterility of cultures that require media supplementation.