Imprimer
Leukotriene E4 ELISA Kit
Marques : Cayman Chemical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
501060-480 501060-96
CAYM501060-480EA 2220 EUR
CAYM501060-480 CAYM501060-96
Leukotriene E4 ELISA Kit
Tests Tests ELISA
For the quantification of LTE4 in urine.
  • Measure LTE4 in urine, with no purification required
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure LTE4 levels down to 25 pg/ml
  • Incubation: 18 hours, Development: 90 to 120 minutes, Read: Colorimetric at 405 to 420 nm
Leukotriene E4 (LTE4) is a product of the 5-lipoxygenase (5-LO) pathway in activated mast cells, eosinophils, and monocytes. LTA4, the primary 5-LO metabolite, is rapidly converted to LTC4 and sequentially to LTD4 and LTE4. While plasma levels of LTC4 are virtually undetectable, exogenously administered LTC4 is recovered in the urine as LTE4 (5 to 13%) and two prominent oxidized metabolites resulting from several cycles of β-oxidation. Cayman’s LTE4 ELISA Kit is a competitive assay developed for the measurement of LTE4 from urine without a purification step. The assay has a range of 7,8 to 1000 pg/ml and a sensitivity (80% B/B0) of approximately 25 pg/ml. This ‘dilute and go’ format has been used successfully to demonstrate significant and reproducible changes in LTE4 levels in the urine of asthmatic patients after antigen challenge. If your samples require additional purification prior to analysis, Cayman’s Cysteinyl Leukotriene Affinity Sorbent can be used for this purpose.
Contents: Antiserum, Tracer, Standard, Buffer, Polysorbate 20, Reagent, Dyes, Plate, and Cover Sheet