Imprimer
MILES BOA® ESD Safety Boots, Closure, Mid, S3
Fournisseur: ELTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
769491/40 769491/41 769491/42 769491/43 769491/44 769491/45 769491/46 769491/47 769491/48
ELTN769491/40EA 132.9 EUR
ELTN769491/40 ELTN769491/41 ELTN769491/42 ELTN769491/43 ELTN769491/44 ELTN769491/45 ELTN769491/46 ELTN769491/47 ELTN769491/48
MILES BOA® ESD Safety Boots, Closure, Mid, S3
Chaussures et bottes
Safety laced boots, MILES, feature the BOA® closure system and are available in black finish.
  • Cowhide leather/Hydrophobised nubuck leather
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • ESD soft-fleece insole with integrated omissions
  • Steel midsole;TPU/PU sole WELLMAXX
  • Abrasion-proof toe protection
  • Composite toe cap
Safety boots feature a fabric lining for natural ventilation.
Certifications: EN ISO 20345 S3 SRC, form B