Imprimer
eStain™ L1 Protein Staining Device
Fournisseur: Genscript
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L00658 L00660 L00661-10 L00662-10 L00663
GENSL00658EA 603 EUR
GENSL00658 GENSL00660 GENSL00661-10 GENSL00662-10 GENSL00663
eStain™ L1 Protein Staining Device
Boîtes de coloration
The eStain™ is a fully automated protein PAGE gel staining system, which uses Coomassie Brilliant Blue and a patented protein staining technology. The system integrates the traditional three steps of fixing, staining, and destaining into one automated process. It can stain and destain two protein PAGE gels simultaneously in 10 minutes.
  • Automated 10-minute staining and destaining for 2 gels simultaneously
  • Consistent results with coomassie blue staining (offered by GenScript) or membrane staining with Ponceau S solution
  • Easy to use with clean setup and convenient operation
  • Sharply stained protein bands with sensitivity ≥12,5 ng
The eStain™ system works with all types of precast as well as homemade PAGE gels. Compared with the conventional staining methods, gels stained by eStain™ have crisp blue bands with minimum or no background. The eStain™ also offers superior sensitivity compared to conventional methods and can detect as low as 12,5ng of protein. The shortened workflow increases productivity. This protein staining system combines three steps of traditional Coomassie blue staining method into one single step to greatly reduce the time required for protein staining.
Excellent uniformity and repeatability with either Coomasie Blue and Ponceau S solution.
Informations de commande: Each kit contains 1×2 L of staining solution, 1×2 L of destaining solution, and a pack of filter paper (50 pK).